ContactUsLinkComponent
CO₂, humidity and temperature data loggers
CO₂, humidity and temperature data loggers
Certificates
undefined
CO₂, humidity and temperature data loggers
Specifications
  • Catalog No:
  • 620-2321
  • 620-2320
  • Interface:
  • USB
  • USB
  • Battery life:
  • 400 days
  • 400 days
  • Weight:
  • 290 g
  • 200 g
  • W×D×H:
  • 234×77×42 mm
  • 120×45×157 mm
  • Data memory - No. of readings:
  • 18000
  • 40000