ContactUsLinkComponent
VWR®, Glass Beads for Grinding and Homogenisation
VWR®, Glass Beads for Grinding and Homogenisation
Catalog # 432-0064
Supplier:  VWR Collection
   Waste Reducing
VWR®, Glass Beads for Grinding and Homogenisation
Catalog # 432-0064
Supplier:  VWR Collection
   Waste Reducing
Specifications
  • Ø:
    0,4 mm
  • Description:
    Silica beads, molecular biology grade
  • Environmentally Preferable:
    Waste Reducing
  • Pk:
    200 g
Specifications
About this item
Small beads suitable for use with vortex mixers and tube homogenisers.
Molecular biology grade beads are treated to be free if DNase and RNase, making them suitable for a range of applications including disruption of mold and yeasts.