SUPELCO Vial Racks
Supplier: Merck
Certificates
About this item
PP, red.
- Can be stacked when empty or full
- Autoclavable 120 °C
- Very good chemical resistance
Economical and convenient storage for samples in screw cap and crimp seal vials.
Frequently Bought Together