ContactUsLinkComponent
Clamps for standard taper joints, WHEATON®
  WHEA297755
undefined
Clamps for standard taper joints, WHEATON®
  WHEA297755
  • For NS:
    29
  • Colour:
    Red
  • Seq:
    02
  • Pk:
    12
These colour-coded clamps moulded from polyoxymethylene (POM), an acetal resin, hold standard taper joints securely under moderate pressure.
  • Will not scratch glass and can withstand temperatures up to 150 °C
  • Resistant to bases and weak acids
  • Not resistant to halogen gases and concentrated acids