Order Entry
Czech Republic
ContactUsLinkComponent
 
 
 
Isoamyl alcohol (mixture of isomers) ≥98%, BAKER ANALYZED®, J.T.Baker®
  9032-01
 :  Avantor
 :  
undefined
 
 
Isoamyl alcohol (mixture of isomers) ≥98%, BAKER ANALYZED®, J.T.Baker®
  9032-01
 :  Avantor
 :  

 

  • Pack type:
    Glass bottle, amber, narrow mouth
  • Pk:
    500 mL
  • Seq:
    2725

 

 

  • For Laboratory,Research,or Manufacturing Use:
    ≥ 98 %
  • Assay (C₅H₁₁OH) (by GC)(total isomers):
    ≥98 %
  • n-Amyl Alcohol (by GC), For Information Only:
    ≤ 0.01 %
  • iso-Amyl Alcohol (by GC), For Information Only:
    ≤ 0.3 %
  • Color (APHA):
    ≤15
  • Acidity (as CH₃COOH):
    ≤0.01 %
  • Water (H₂O)(by Karl Fischer titrn):
    ≤0.3 %
  • Residue after Evaporation:
    ≤0.0050 %
  • Density (g/mL) at 20°C:
    0.812 - 0.819
  • Aldehydes:
    Passes Test

 

 

 :
 :88.15 g/mol
 :13,07195
 :137.9 °C (1013 hPa)
 :–79 °C
 :0.81 g/cm³ (20 °C)
 :48 °C
 :Ambient
 :200-752-1
 :1105
 :3,III
 :GHS02
 :GHS07
 :30899-19-5
 :C₅H₁₂O
 :MFCD00002977