Order Entry
Czech Republic
ContactUsLinkComponent
Potassium hexacyanoferrate(III) ≥99.0% (by iodometry), crystals, BAKER ANALYZED® ACS, J.T.Baker®
Potassium hexacyanoferrate(III) ≥99.0% (by iodometry), crystals, BAKER ANALYZED® ACS, J.T.Baker®
Supplier:  Avantor
Certificates
undefined
Potassium hexacyanoferrate(III) ≥99.0% (by iodometry), crystals, BAKER ANALYZED® ACS, J.T.Baker®
Supplier:  Avantor
Certificates

Specification Test Results

  • Meets ACS Reagent Chemical Requirements:
    ≥99.0%
  • For Laboratory,Research,or Manufacturing Use:
    ≤0.005%
  • Appearance:
    Passes Test
  • Assay (K₃Fe(CN)₆) (by iodometry):
    ≥99.0 %
  • Insoluble Matter:
    ≤0.005 %
  • Chloride (Cl):
    ≤0.01 %
  • Sulfate (SO₄):
    ≤0.01 %
  • Ferro Compounds (as [Fe(CN)₆]⁴⁻):
    ≤0.05 %

Specification Test Results

Related Information

Frequently Bought Together