ContactUsLinkComponent
Swab holder, SwabTool™
Swab holder, SwabTool™
Catalog # 300-0236
Supplier:  MICRONOVA
undefined
Swab holder, SwabTool™
Catalog # 300-0236
Supplier:  MICRONOVA
Specifications
  • Description:
    SwabTool™ adapter, 304 electropolished stainless steel; acetal components
  • Seq:
    05
  • Pk:
    1
Specifications
About this item
A custom tool specifically designed to hold swabs for environmental monitoring and sampling. Use with SSU series handles for extension in large tanks and other hard to reach areas.
  • Swab is locked in place with a twist knob
  • Adapted to swabs with a 6,4 mm handle
  • Optional 'straws' available to hold smaller or longer swabs
  • Crafted from electropolished stainless steel with acetal components
  • Quick Disconnect fitting connects to SSU series stainless steel handles
  • Compatible with strong disinfectants
  • Autoclavable
The Micronova SwabTool™ Adapter is compatible with a range of swabs when used with optional 'straws' to hold those which are smaller or longer. Choose from the range of SSU series handles to match the length required for reach.