ContactUsLinkComponent
Examination gloves, latex, Unigloves® Safetec
Examination gloves, latex, Unigloves® Safetec
Certificates
undefined
Examination gloves, latex, Unigloves® Safetec
Specifications
  • Catalog No:
  • 112-1782
  • 112-1779
  • 112-1780
  • 112-1781
  • Model:
  • Safetec
  • Safetec
  • Safetec
  • Safetec
  • Ambidextrous / hand-specific:
  • Ambidextrous
  • Ambidextrous
  • Ambidextrous
  • Ambidextrous
  • Disposable / reusable:
  • Disposable
  • Disposable
  • Disposable
  • Disposable
  • Powdered / powder-free:
  • Powder-free
  • Powder-free
  • Powder-free
  • Powder-free
  • Finish:
  • Micro-textured
  • Micro-textured
  • Micro-textured
  • Micro-textured
  • Cuff style:
  • Beaded
  • Beaded
  • Beaded
  • Beaded
  • Cuff thickness:
  • 0,11 - 0,15 mm
  • 0,11 - 0,15 mm
  • 0,11 - 0,15 mm
  • 0,11 - 0,15 mm
  • Finger thickness:
  • 0,18 - 0,22 mm
  • 0,18 - 0,22 mm
  • 0,18 - 0,22 mm
  • 0,18 - 0,22 mm
  • Palm thickness:
  • 0,15 - 0,18 mm
  • 0,15 - 0,18 mm
  • 0,15 - 0,18 mm
  • 0,15 - 0,18 mm
  • EN norm:
  • EN 455
  • EN 455
  • EN 455
  • EN 455