

Hydrochloric acid 36.5 - 38.0%, AR® ACS, Macron Fine Chemicals™
Katalogové číslo H613-27
Dodavatel: MACRON AVANTOR BRAND
Číslo CAS:
Specifikace
- Pack type:PE drum, black
- Pk:500 lb
- Seq:7323
Specifikace
Výsledky testů specifikací
- Meets ACS Reagent Chemical Requirements:—
- For Laboratory,Research,or Manufacturing Use:—
- ACS - Assay (HCl):36.5 - 38.0 %
- Appearance:Passes Test
- ACS - Color (APHA):≤ 10
- ACS - Ammonium (NH₄):≤ 3 ppm
- ACS - Arsenic (As):≤ 0.005 ppm
- ACS - Bromide (Br):≤ 0.005 %
- ACS - Extractable Organic Substances:≤ 5 ppm
- ACS - Free Chlorine (as Cl):≤ 1 ppm
- ACS - Heavy Metals (as Pb):≤ 0.1 ppm
- ACS - Residue after Ignition:≤ 4 ppm
- ACS - Sulfate (SO₄):≤ 0.5 ppm
- ACS - Sulfite (SO₃):≤ 0.8 ppm
- Trace Impurities - Aluminum (Al):≤ 0.300 ppm
- Trace Impurities - Boron (B):≤ 0.100 ppm
- Trace Impurities - Chromium (Cr):≤ 0.200 ppm
- Trace Impurities - Copper (Cu):≤ 0.10 ppm
- Trace Impurities - Gold (Au):≤ 0.100 ppm
- Trace Impurities - Iron (Fe):≤ 0.1 ppm
- Trace Impurities - Lead (Pb):≤ 0.100 ppm
- Trace Impurities - Magnesium (Mg):≤ 0.300 ppm
- Trace Impurities - Manganese (Mn):≤ 0.300 ppm
- Trace Impurities - Nickel (Ni):≤ 0.1 ppm
- Trace Impurities - Potassium (K):≤ 0.300 ppm
- Trace Impurities - Tin (Sn):≤ 0.300 ppm
- Trace Impurities - Titanium (Ti):≤ 0.300 ppm
- Trace Impurities - Zinc (Zn):≤ 0.100 ppm
Výsledky testů specifikací
O tomto produktu
:
Bod varu:110 °C (1013 hPa)
Bod tání:–30 °C
Hustota:1.18 g/cm³ (20 °C)
Teplota pro skladování:Ambient
UN:1789
ADR:8,II
:Management of Change (MOC) category = N
:36.46 g/mol
EINECS:231-595-7
:GHS05
:GHS07
Číslo CAS:7647-01-0
Vzorec:HCl
MDL číslo:MFCD00011324