Order Entry
Czech Republic
ContactUsLinkComponent
 
Florisil® 100 - 200 mesh, J.T.Baker®
Catalog # M369-07
Supplier:  Avantor
CAS Number:  
undefined
Florisil® 100 - 200 mesh, J.T.Baker®
Catalog # M369-07
Supplier:  Avantor
CAS Number:  

Specifications

  • Pack type:
    Poly bottle, white, blue T/E cap
  • Pk:
    500 g
  • Seq:
    3645

Specifications

About this item

 :
 :100.38 g/mol
Density:2.51 Kg/L
EINECS:215-681-1
CAS Number:1343-88-0
Formula:MgO₃Si
MDL Number:MFCD00078249