Destaining solution, Blue-OUT™ for mass spectrometry
Catalog # 786-683
Supplier: G-Biosciences
CAS Number:
Specifications
- Pk:1 KIT
- Seq:0075
- Pk Info:10 ml (100 preps)
Specifications
About this item
:
Storage Temperature:Ambient
UN:1648
ADR:3,II
:GHS02
:GHS06