Order Entry
Czech Republic
Orders LinkContactUsLinkComponent
Mortars with pestles

Mortars with pestles

1000 ml, 80 mm, 150 mm
Catalog # NEUB3420-1002
Mortars with pestles
Catalog # NEUB3420-1002
Mortars with pestles

1000 ml, 80 mm, 150 mm

Price
3 320,00 Kč
Log in for availability
1 * 1 KS
Product Details & Documents

Glass, inner surfaces frosted glass.

Specifications
  • Ø:
    150 mm
  • Capacity:
    1000 ml
  • Height:
    80 mm