ContactUsLinkComponent
VWR®, Immersion Oil
VWR®, Immersion Oil
Catalog # 630-1808
Supplier:  VWR Collection
CAS Number:  
undefined
VWR®, Immersion Oil
Catalog # 630-1808
Supplier:  VWR Collection
CAS Number:  
Specifications
  • Pk:
    1
  • Description:
    Immersion oil (8 ml)
Specifications
About this item
Immersion oil is used to increase microscope resolution.
  • For immersing of the lens and specimen
  • Transparent oil with high refractive index
  • Increases the numerical aperture of an objective