ContactUsLinkComponent
CytoScan™ Cell proliferation assay kit, WST-1
CytoScan™ Cell proliferation assay kit, WST-1
Catalog # 786-212
Supplier:  G-Biosciences
CAS Number:  
undefined
CytoScan™ Cell proliferation assay kit, WST-1
Catalog # 786-212
Supplier:  G-Biosciences
CAS Number:  
Specifications
  • Pk:
    500 Assays
  • Description:
    CytoScan™ WST-1 Cell Proliferation Assay Kit
  • Tests per kit:
    500 assays
Specifications
About this item
CytoScan™ WST-1 Cell Proliferation Assay principle is based upon the reduction of the tetrazolium salt WST-1 to formazan by cellular dehydrogenases. The generation of the dark yellow coloured formazan is measured at 420 to 480 nm (optimal at 440 nm) and is directly correlated to cell number.
  • Sensitive and accurate assay
  • Colorimetric assay
  • No washing, harvesting or solubilization required
  • For the detection of cell toxicity, death, viability or proliferation
  • Ideal for high throughput screening