


Ammonia solution 30%, BAKER INSTRA-ANALYZED® for trace metal analysis, J.T.Baker®
Supplier: Avantor
Certificates
About this item
CAS Number:1336-21-6
:7664-41-7
Formula:NH₃
MDL Number:MFCD00066650
:35.05 g/mol
Boiling Pt:32…37.7 °C (1013 hPa)
Melting Pt:–60…–57.5 °C
Density:0.88…0.91 g/cm³ (20 °C)
EINECS:215-647-6
UN:2672
ADR:8,III
:GHS05
:GHS07
:GHS09
Storage Temperature:Ambient
Specification Test Results
- For Laboratory,Research,or Manufacturing Use:28.0 - 30.0 %
- Assay (as NH₃):28.0 - 30.0 %
- Specific Gravity at 60°/60°F:0.896 - 0.902
- Appearance:Passes Test
- Color (APHA):≤10
- Residue after Ignition:≤3 ppm
- Substances Reducing Permanganate:Passes Test
- Chloride (Cl):≤0.5 ppm
- Phosphate (PO₄):≤0.4 ppm
- Total Sulfur (as SO₄):≤1 ppm
- Trace Impurities - Aluminum (Al):≤20 ppb
- Trace Impurities - Arsenic and Antimony (as As):≤50.0 ppb
- Trace Impurities - Barium (Ba):≤10.0 ppb
- Trace Impurities - Boron (B):≤20.0 ppb
- Trace Impurities - Cadmium (Cd):≤5.0 ppb
- Trace Impurities - Calcium (Ca):≤200.0 ppb
- Trace Impurities - Chromium (Cr):≤5.0 ppb
- Trace Impurities - Cobalt (Co):≤1.0 ppb
- Trace Impurities - Copper (Cu):≤20.0 ppb
- Heavy Metals (as Pb):≤200.0 ppb
- Trace Impurities - Iron (Fe):≤10.0 ppb
- Trace Impurities - Lead (Pb):≤5.0 ppb
- Trace Impurities - Lithium (Li):≤20.0 ppb
- Trace Impurities - Magnesium (Mg):≤20 ppb
- Trace Impurities - Manganese (Mn):≤5.0 ppb
- Trace Impurities - Mercury (Hg):≤5.0 ppb
- Trace Impurities - Nickel (Ni):≤5.0 ppb
- Trace Impurities - Potassium (K):≤500.0 ppb
- Trace Impurities - Silicon (Si):≤1000.0 ppb
- Trace Impurities - Silver (Ag):≤5.0 ppb
- Trace Impurities - Sodium (Na):≤300 ppb
- Trace Impurities - Strontium (Sr):≤10.0 ppb
- Trace Impurities - Tin (Sn):≤10.0 ppb
- Trace Impurities - Zinc (Zn):≤5.0 ppb
Specification Test Results
Frequently Bought Together