

Acetic acid glacial ≥99.9%, BAKER INSTRA-ANALYZED® for trace metal analysis, J.T.Baker®
Supplier: Avantor
Certificates
About this item
CAS Number:64-19-7
:
Formula:H₃CCOOH
MDL Number:MFCD00036152
:60.05 g/mol
Boiling Pt:118 °C (1013 hPa)
Melting Pt:17 °C
Density:1.05 g/cm³ (20 °C)
Flash Pt:38.5 °C
Storage Temperature:Ambient
EINECS:200-580-7
UN:2789
ADR:8,II
REACH:01-2119475328-30
:GHS02
:GHS05
Specification Test Results
- For Laboratory,Research,or Manufacturing Use:= 99.9 %
- IMPORTANT: Material will freeze if stored below 17 °C (63°F).:= 0.005 %
- ACS - Assay (by GC, corrected for water):≥ 99.9 %
- ACS - Acetaldehyde:≤ 0.005 %
- ACS - Acetic Anhydride ((CH₃CO)₂O):≤ 0.01 %
- ACS - Color (APHA):≤ 10
- Dilution Test:Passes Test
- ACS - Residue after Evaporation:≤ 5 ppm
- ACS - Solubility in H₂O:Passes Test
- ACS - Specific Gravity at 20°/20°C:≥ 1.048
- ACS - Substances Reducing Dichromate:Passes Test
- ACS - Substances Reducing Permanganate:Passes Test
- ACS - Titrable Base (meq/g):≤ 0.0004
- Chloride (Cl):≤ 0.5 ppm
- Phosphate (PO₄):≤ 1 ppm
- Sulfate (SO₄):≤ 0.5 ppm
- Trace Impurities - Aluminum (Al):≤ 50.0 ppb
- Arsenic and Antimony (as As):≤ 5.0 ppb
- Trace Impurities - Barium (Ba):≤ 20.0 ppb
- Trace Impurities - Beryllium (Be):≤ 10.0 ppb
- Trace Impurities - Bismuth (Bi):≤ 50 ppb
- Trace Impurities - Boron (B):≤ 10.0 ppb
- Trace Impurities - Cadmium (Cd):≤ 5.0 ppb
- Trace Impurities - Calcium (Ca):≤ 100.0 ppb
- Trace Impurities - Chromium (Cr):≤ 50 ppb
- Trace Impurities - Cobalt (Co):≤ 10.0 ppb
- Trace Impurities - Copper (Cu):≤ 10.0 ppb
- Trace Impurities - Germanium (Ge):≤ 50.0 ppb
- Heavy Metals (as Pb):≤ 300.0 ppb
- Trace Impurities - Iron (Fe):≤ 50.0 ppb
- Trace Impurities - Lead (Pb):≤ 10.0 ppb
- Trace Impurities - Lithium (Li):≤ 50.0 ppb
- Trace Impurities - Magnesium (Mg):≤ 50.0 ppb
- Trace Impurities - Manganese (Mn):≤ 5.0 ppb
- Trace Impurities - Mercury (Hg):≤ 5.0 ppb
- Trace Impurities - Nickel (Ni):≤ 10.0 ppb
- Trace Impurities - Potassium (K):≤ 200.0 ppb
- Trace Impurities - Silicon (Si):≤ 50 ppb
- Trace Impurities - Silver (Ag):≤ 5.0 ppb
- Trace Impurities - Sodium (Na):≤ 500.0 ppb
- Trace Impurities - Strontium (Sr):≤ 5.0 ppb
- Trace Impurities - Tin (Sn):≤ 10.0 ppb
- Trace Impurities - Titanium (Ti):≤ 300.0 ppb
- Trace Impurities - Zinc (Zn):≤ 10.0 ppb
Specification Test Results
Frequently Bought Together