Order Entry
Czech Republic
ContactUsLinkComponent
VWR®, Beard Cover, PP
VWR®, Beard Cover, PP
Catalog # 113-7920
Supplier:  VWR Collection
undefined
VWR®, Beard Cover, PP
Catalog # 113-7920
Supplier:  VWR Collection

Specifications

  • Colour:
    White
  • Pk:
    1.000
  • Weight:
    11 g/m²

Specifications

About this item

Basic beard cover made from non woven PP. For non critical applications.

  • Elasticated around the neck
  • Breathable, soft material
  • Latex-free

Size: 46 cm
Weight: 11 g/m²