Order Entry
Czech Republic
ContactUsLinkComponent
2-Iminothiolane hydrochloride (Traut's Reagent)
2-Iminothiolane hydrochloride (Traut's Reagent)
Catalog # BC95
Supplier:  G-Biosciences
CAS Number:  
undefined
2-Iminothiolane hydrochloride (Traut's Reagent)
Catalog # BC95
Supplier:  G-Biosciences
CAS Number:  

Specifications

  • Pk:
    500 mg
  • Seq:
    2725

Specifications

About this item

 :
 :137.625 g/mol
CAS Number:4781-83-3
Formula:C₄H₈ClNS
MDL Number:MFCD00039013