ContactUsLinkComponent
FASTsilver™, rapid silver stain for protein gels and nucleic acids
Catalog # 786-30
Supplier:  G-Biosciences
CAS Number:  
undefined
FASTsilver™, rapid silver stain for protein gels and nucleic acids
Catalog # 786-30
Supplier:  G-Biosciences
CAS Number:  
Specifications
  • Pk:
    1 KIT
  • Description:
    FASTsilver™ kit
  • Pk Info:
    25 Mini Gels
Specifications
About this item
G-Biosciences' FASTsilver™ is a rapid silver stain for nucleic acids and proteins in electrophoresis gels
  • Nanogram-sensitive
  • Stains both nucleic acids & proteins
  • Maximum visibility: produces crystal clear backgrounds & sharp protein bands
  • Lower Detection Limit: 1ng/band
FASTsilver™ does not use the protein modifier glutaraldehyde, and therefore, it allows for complete recovery of proteins and trypsin digested peptides.
G-Biosciences' FASTsilver™ detects as little as 0.3 ng nucleic acid and 1 ng BSA protein. The kit contains ready-to-use reagents for 25 mini-gels and comes with a simple-to-follow 60 to 90 minute protocol.
The FASTsilver™ kit is a nanogram-sensitive silver staining kit that produces crystal clear background and maximal sensitivity needed for critical analysis. This unique formulation of reagents leaves the background clear and produces sharp images of nucleic acid bands.