ContactUsLinkComponent
Protein concentration kit, OrgoSol-PROTEIN-Concentrate™
Protein concentration kit, OrgoSol-PROTEIN-Concentrate™
Catalog # 786-125
Supplier:  G-Biosciences
CAS Number:  
undefined
Protein concentration kit, OrgoSol-PROTEIN-Concentrate™
Catalog # 786-125
Supplier:  G-Biosciences
CAS Number:  
Specifications
  • Pk:
    50 mL
  • Description:
    OrgoSol-PROTEIN-Concentrate™ Kit, for 50 ml
Specifications
About this item
The OrgoSol-PROTEIN-Concentration™ Kit precipitates protein with a proprietary solvent buffer, OrgoSol™. The OrgoSol™ buffer has been specifically developed for efficient precipitation of protein solutions with minimal disruption to the protein structure and therefore maintains the biological activity of most proteins.
The kit has been extensively tested for the concentration of a wide selection of enzymatic proteins without the loss of their biological activity and for ~100% protein recovery. The kit is designed to precipitate up to 5 ml protein solution.
The method involves mixing a protein solution with the OrgoSol™ Buffer followed by incubation, which results in quantitative precipitation of the protein. The precipitated protein is suspended in a smaller volume of an appropriate buffer and the concentrated protein solution is ready for use.