ContactUsLinkComponent
Protease Screening Kit
Protease Screening Kit
Catalog # 786-137
Supplier:  G-Biosciences
CAS Number:  
Protease Screening Kit
Catalog # 786-137
Supplier:  G-Biosciences
CAS Number:  
Specifications
  • Pk:
    1 KIT
  • Description:
    Protease Screening Kit
  • Tests per kit:
    50 assays
Specifications
About this item
The Protease Screening Kit provides a simple and quick method for testing samples for proteolysis. Simply incubate sample in the reagent provided and obtain results.
The kit uses dye-labelled protein conjugate as protease substrate, which allows nanogram level detection. The absorbance of dye-labelled peptide is measured at 574 nm for determination of protease activity.
The kit is sufficient for 50 assays in a microwell format.