ContactUsLinkComponent
Protease inhibitor cocktail, FOCUS™ ProteaseArrest™
Protease inhibitor cocktail, FOCUS™ ProteaseArrest™
Catalog # 733-1945
Supplier:  G-Biosciences
CAS Number:  
undefined
Protease inhibitor cocktail, FOCUS™ ProteaseArrest™
Catalog # 733-1945
Supplier:  G-Biosciences
CAS Number:  
Specifications
  • Pk:
    1 mL
  • Description:
    FOCUS™ ProteaseArrest™ (100X)
Specifications
About this item
A ready to use, 100X concentrated, broad range protease inhibitor cocktail that is fully compatible with 2D electrophoresis and subsequent mass spectrometry.
  • Compatible to 2D gel electrophoresis
  • Broad spectrum protease inhibitor cocktail
  • 100X concentrated, ready to use solution
  • Inhibition of protease activity in protein preparations
  • Suitable for 2D gel sample preparation for protection of protein samples