ContactUsLinkComponent
Total protein extraction kit, TPE™
Total protein extraction kit, TPE™
Catalog # 786-225
Supplier:  G-Biosciences
CAS Number:  
undefined
Total protein extraction kit, TPE™
Catalog # 786-225
Supplier:  G-Biosciences
CAS Number:  
Specifications
  • Pk:
    1 KIT
  • Description:
    Total Protein Extraction (TPE™) Kit
Specifications
About this item
Total Protein Extraction (TPE™) Kit is a universal lysis system for the solubilisation of total proteins from animal, plant, yeast, bacteria, and other biological samples. The kit contains ready to use buffers for extracting total protein from cells and tissues for SDS-PAGE analysis.
The TPE™ Kit provides a two component protocol that eliminates clump formation, protein loss, and other problems associated with total protein extraction procedures. Samples are ground in the buffer provided and then heated to solubilize the total protein.
The TPE™ Kit is based on optimised concentrations of Tris and SDS and is suitable for running denaturing electrophoresis and other downstream applications. The kit is supplied with sufficient reagents for 50×250 mg preparations.