Vacuum jars, Nalgene®
Supplier: Thermo Fisher Scientific
Certificates
About this item
Polyetherimide, amber, transparent.
- Excellent chemical resistance (not use with unsaturated halogenated hydrocarbons)
- Good temperature resistance
- Not autoclavable when used for vacuum service
These chemically-resistant jars have an amber cast and may be useful for demanding service.
Specifications
- Catalog No:
- NALGDS5320-1212
- NALGDS5320-0910
- Environmentally Preferable:
- Low Manufacturing Impact
- Low Manufacturing Impact
- Environmental certification:
- NAR Certificate of Retirement
- IRECS; ISO 14001 (Certificate Registration No. EMS 752603)