Cartridge filters, Fluorogard® FP
Supplier: ENTEGRIS
Certificates
About this item
These filters can be used in solvent based photochemicals without the need for prewetting steps.
- Excellent chemical compatibility
- For use in POU filtration and chemical delivery system filtration
Membrane: Hydrophobic PTFE; Supports, core, sleeve and endcaps: PP
Maximum operating temperature: 90 °C
Maximum forward/reverse differential pressure: 3,4 bar at 25 °C
Frequently Bought Together