Order Entry
Czech Republic
ContactUsLinkComponent
Drums, wide neck, with screw cap and tap
Drums, wide neck, with screw cap and tap
  DELT209030
 :  DELTALAB
undefined
Drums, wide neck, with screw cap and tap
  DELT209030
 :  DELTALAB

 

  • Ø opening:
    150 mm
  • Ext.Ø:
    320 mm
  • Capacity:
    30 L
  • Height:
    470 mm
  • Pk:
    1

 

 

PE, wide neck, with green PE screw cap.

  • Cap with non hermetical plug
  • With a tap and two black folding handles at the top
  • Not designed to transport liquids