Glass wool, Supelco®
: Merck
Wool plugs are often used in GC inlet liners to improve sample vaporization, and/or to keep non-volatile material from entering the column. They can also be used in packed GC columns, solvent desorption tubes, thermal desorption tubes, and purge traps to retain packed beds.
Silanised: For general purpose use
Pesticide Grade (Silanised): For applications that involve active analytes, such as organochlorine and/or organophosphorous pesticides.
Phosphoric acid treated: For acidic compounds such as barbiturates, free fatty acids, etc.
Non-treated: For special application where the user deactivates based on their intended analytes
Frequently Bought Together