ContactUsLinkComponent
Fisher Container™ Autoclavable Precision Clean® Breather Bags
Fisher Container™ Autoclavable Precision Clean® Breather Bags
Catalog # FISH90726P
Supplier:  FISHER CONTAINER
undefined
Fisher Container™ Autoclavable Precision Clean® Breather Bags
Catalog # FISH90726P
Supplier:  FISHER CONTAINER
Specifications
  • Description:
    Autoclavable Precision Clean® Breather Bags
  • L×W:
    210×310 mm
  • Pk:
    500
Specifications
About this item
A combination of Tyvek™ and clear, ultraclean film structure aids in the prevention of interior particle contamination. Designed for products to be used in cleanroom environments that also require sterilization.
  • Faster cycle times
  • Molecular bonded seals
  • Pouch suitable for sterilizaiton by steam, EtO, gamma radiation or hydrogen peroxide
  • Tyvek™ 1059B one side and one side 3Mil PCHD
Certifications: Certified to NASA™ clean standards by Fisher to meet Level 100 KSC-C-123