0
0,00 Kč
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Czech Republic
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Block Heaters
Digital Block Heaters
Accessories for digital block reactor, DRB 200
Accessories for digital block reactor, DRB 200
Accessories for digital block reactor, DRB 200
Catalog # HACHHHA155
Supplier:
Hach
Expand image
Accessories for digital block reactor, DRB 200
Catalog # HACHHHA155
Supplier:
Hach
Specifications
Description:
Reducing adapter for DRB200, converts 20 mm diameter vial wells to 16 mm diameter
Pk:
1
Specifications
About this item
Product Details
Product Details