ContactUsLinkComponent
Cleanroom wipes, C-Wipes, C6
Cleanroom wipes, C-Wipes, C6
Catalog # EPTA7-C6-99L-00
Cleanroom wipes, C-Wipes, C6
Catalog # EPTA7-C6-99L-00
Specifications
  • Packed:
    500/bag
  • L×W:
    230×230 mm
  • Pk:
    500
Specifications
About this item
This lightweight, low linting tissue is made from a specialty blend of hemp and cellulose fibers. It is ideal for general purpose cleaning applications.
  • Effective in the cleanroom because of their super absorbency, extremely low particle generation, and ultra purity
  • Absorb four times their own weight
  • Soft wipes will not scratch surfaces when used either wet or dry
  • Low in soluble extractables and metallic ions
Colour: White
Packaging: Converted, cleaned, and double-bag packaged in a cleanroom environment.