Cut and flame resistant gloves, ActivArmr® 80-813
: Ansell
These knitted gloves combine outstanding flame resistance, high cut protection and ergonomic design. The exclusive composite yarn made of Modacrylic, glass fiber and Kevlar® provides exceptional cut resistance for safe handling of sharp objects and materials.
- Materials used in the glove are inherently flame resistant, even after washing
- Arc Flash Hazard/Risk Category 2: helps protect workers against arc flash hazards
- Soft foam neoprene coating on the palm to ensure great flexibility and a secure grip in a variety of environments
- Ergonomic design for superior comfort and reduced hand fatigue
- Knitwrist cuff
- Antistatic according to EN 1149
Suitable applications include: Equipment assembly, replacing filters and parts, facility and machinery maintenance, pipe installation and repair, carpentry/scaffolding and heavy duty metal handling.
Gauge: 13
Colour: Black/green
EN 388 (2542), EN 407 (412110); Cat. III
: EN 388:2016 (2X42C), EN 407 (412110); Cat. III
Arc Flash - Test Method/Standard: ASTM F2675-13 - Result: Hazard/Risk Category (HRC) 2 - Arc Rating (ATPV) 9,4 cal/cm².
Arc Flash - Test Method/Standard: ASTM F2675-13 - Result: Hazard/Risk Category (HRC) 2 - Arc Rating (ATPV) 9,4 cal/cm².
- Catalog No:
- ANSE80-813/8
- ANSE80-813/7
- ANSE80-813/6
- ANSE80-813/11
- ANSE80-813/10
- ANSE80-813/9
- Model:
- 80-813
- 80-813
- 80-813
- 80-813
- 80-813
- 80-813
- Ambidextrous / hand-specific:
- Hand-specific
- Hand-specific
- Hand-specific
- Hand-specific
- Hand-specific
- Hand-specific
- Disposable / reusable:
- Reusable
- Reusable
- Reusable
- Reusable
- Reusable
- Reusable
- Liner material:
- Modacrylic/glass fibre/Kevlar®
- Modacrylic/glass fibre/Kevlar®
- Modacrylic/glass fibre/Kevlar®
- Modacrylic/glass fibre/Kevlar®
- Modacrylic/glass fibre/Kevlar®
- Modacrylic/glass fibre/Kevlar®
- Finish:
- Palm coated
- Palm coated
- Palm coated
- Palm coated
- Palm coated
- Palm coated
- Gauge:
- 13
- 13
- 13
- 13
- 13
- 13
- Cuff style:
- Knit wrist
- Knit wrist
- Knit wrist
- Knit wrist
- Knit wrist
- Knit wrist
- EN norm:
- EN 388:2016 (2X42C), EN 407 (412110)
- EN 388:2016 (2X42C), EN 407 (412110)
- EN 388:2016 (2X42C), EN 407 (412110)
- EN 388:2016 (2X42C), EN 407 (412110)
- EN 388:2016 (2X42C), EN 407 (412110)
- EN 388:2016 (2X42C), EN 407 (412110)
- Anti-static:
- +
- +
- +
- +
- +
- +
Frequently Bought Together