ContactUsLinkComponent
Multipurpose basins, with handles
  SEMA1752
 :  SEMADENI
Multipurpose basins, with handles
  SEMA1752
 :  SEMADENI
  • Capacity:
    11,0 l
  • Colour:
    Grey
  • W×D×H:
    390×300×150 mm
  • Pk:
    1
PP, granite.
  • Rectangular, with two side handles
  • Stackable and resistant in hot water
  • Suitable for food
Multipurpose basins for a wide range of applications.