ContactUsLinkComponent
Acrodisc® Syringe Filters, WBC (White Blood Cell)
Acrodisc® Syringe Filters, WBC (White Blood Cell)
   Sustainable Options Available
Certificates
undefined
Acrodisc® Syringe Filters, WBC (White Blood Cell)
   Sustainable Options Available
Specifications
  • Catalog No:
  • PALLAP-4951
  • PALLAP-4952
  • Environmentally Preferable:
  • Waste Reducing
  • Waste Reducing
  • Max. operating pressure:
  • 30 psi
  • 30 psi
  • Max. temperature:
  • 34 °C
  • 34 °C