Multi-use temperature and humidity loggers, testo 184 H1
Catalog # 620-2361
Supplier: Testo
Specifications
- Range:T: −20…+70 °C
RH: 0 - 100 % - Data memory - No. of readings:64000
- Interface:USB
- Description:Mini temperature and humidity logger, 184 H1
- Seq:01
- Battery life:500 days
- Alarm:Visual
- Batteries:Lithium battery CR2450, 3 V, exchangeable
- Display:Segment LCD; one-line
- Weight:44 g
- W×D×H:44×12×97 mm
- Accuracy:T: ±0,5 °C (0…70 °C ); ±0,8 °C (−20…0 °C );
RH: 0,1 % - Pk:1
Specifications
About this item
The 184 H1 temperature and humidity logger is capable of monitoring temperature and humidity levels during cold chain transportation. With configuration, reports, and calibration certificates on-board, the 184 H1 make transport monitoring and documentation of sensitive goods easy.
- PDF reports allow for quick access to non-corruptible documentation without installed software
- On-board electronic calibration certificate eliminates the hassles of maintaining traceability
- Easy configuration on any computer with pre-loaded files, clear alarm indication
- Its large, easy-to-read display shows readouts in real time
- A LED status lights alert when specified limits have been exceeded
Certifications: IP 30 - ISO 9001:2008, FDA 21 CFR Part 11, EN 12830, and HACCP (Hazard Analysis Critical Control Point) International compliant.
Delivery information: Supplied with replaceable long life battery, up to 500 consecutive days of use.