Order Entry
Czech Republic
ContactUsLinkComponent
 
Tape dispenser, stainless steel
Catalog # 817-0311
Supplier:  ULTRATAPE
undefined
Tape dispenser, stainless steel
Catalog # 817-0311
Supplier:  ULTRATAPE

Specifications

  • For Use With:
    Tapes with 51 mm width
  • Colour:
    Grey
  • Description:
    Tape dispenser, stainless steel
  • Pk:
    1

Specifications

About this item

These dispensers are made from industrial 304 grade stainless steel with a mounting bracket on the bottom. It is corrosion resistant and the replaceable serrated blade is electroplated chrome buffed to a dull shine. It is designed to hold one roll, with a 76 mm core, up to 50,8 mm width and 201,2 metres in length.