Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 Kč
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Czech Republic
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
m-Cresol
m-Cresol 15 g/l
m-Cresol 15 g/l
Irritant
m-Cresol 15 g/l
Catalog # VWRS30173.465
Supplier:
VWR Collection
CAS Number:
SDS
Certificates
Expand image
Irritant
m-Cresol 15 g/l
Catalog # VWRS30173.465
Supplier:
VWR Collection
CAS Number:
SDS
Certificates
Specifications
Pk:
25 L
Seq:
5327
Specifications
About this item
Product Details
Documentation
:
:
108.14 g/mol
:
GHS07
CAS Number:
108-39-4
Formula:
CH₃C₆H₄OH
MDL Number:
MFCD00002302
Product Details
Documentation