Order Entry
Czech Republic
ContactUsLinkComponent
 
Pharmacist's spatula
Catalog # BOCH3760
Supplier:  BOCHEM
undefined
Pharmacist's spatula
Catalog # BOCH3760
Supplier:  BOCHEM

Specifications

  • Length:
    230 mm
  • Environmentally Preferable:
  • Blade width:
    20 mm
  • Pk:
    1
  • Blade length:
    130 mm

Specifications

About this item

Pharmacist's spatula with wooden handle and PTFE coated blade.