ContactUsLinkComponent
Microtubes with cap, 1,5 ml
Microtubes with cap, 1,5 ml
Catalog # 211-2081
Supplier:  SEMADENI
undefined
Microtubes with cap, 1,5 ml
Catalog # 211-2081
Supplier:  SEMADENI
Specifications
  • Type:
    with attached lid
  • Version:
    graduated, conical bottom, non sterile
  • Capacity:
    1,5 ml
  • Colour:
    Natural
  • Seq:
    01
  • Pk:
    1.000
Specifications
About this item
PP, translucent.
  • Pierceable lid
  • Autoclavable
  • Withstand centrifugation at 21000×g
  • Excellent chemical resistance
With attached push-on lid.