VWR® kuroGEL, Semi-dry blotter
Catalog # 700-0416
Supplier: VWR Collection
Specifications
- Capacity:4 blots: 80×85 mm
2 blots: 160×85 mm
1 blot: 150×175 mm - Description:Semi-dry blotter, kuroGEL
- Model:kuroGEL semi-dry blotter
- Buffer volume:20 ml
- Unit W×D×H:325×250×55 mm
- Pk:1
Specifications
About this item
The kuroGEL semi-dry blotter provides great flexibility in a laboratory since it can be used for all types of blotting: Western, Southern and Northern. Blotting is fast and easy, and can typically be completed in 15 to 30 min. Set-up procedures are easy and economical with little buffer needed. The screw down lid adjusts to varying gel thicknesses and sizes, while the platinum coated electrodes provide uniform pressure ensuring even transfers.
- Suitable for Western, Southern and Northern blots
- Rapid transfer times
- Uniform heat dispersion
- Variable gel thickness
- Minimal buffer volume
Delivery: Each unit includes base with plate electrode, lid with plate electrode, electrical leads, and 2× unit screws.