ContactUsLinkComponent
Anaerobic gas system, PetriSphere
Anaerobic gas system, PetriSphere
Catalog # 710-1964
Supplier:  BioTool
undefined
Anaerobic gas system, PetriSphere
Catalog # 710-1964
Supplier:  BioTool
Specifications
  • Description:
    Anaerobic gas system, PetriSphere without jar
  • Pk:
    1
Specifications
About this item
The PetriSphere anaerobic gas system is suitable for the culturing of anaerobic, microaerophilic and capnophilic bacteria in common anaerobic jars. Atmospheric conditions can be developed inside the jar in less than one minute, with the flexibility to choose the level of oxygen depletion. Gas extraction can be controlled to reduce risk of cell lysis. The PetriSphere system consists of a microprocessor-based controller, a diaphragm vacuum pump and a gas reservoir connection, which enables the air to be replaced with nitrogen or carbon dioxide gas. By simply pressing the start button the process starts and runs fully automatically.
  • No additional chemicals are needed to create an anaerobic atmosphere
  • Special click-connectors provide convenient, safe and time saving operation
  • Anaerobic atmosphere in less than 3 minutes