ContactUsLinkComponent
Peha-Soft® Nitrile Nitrile Gloves, Sterile
Peha-Soft® Nitrile Nitrile Gloves, Sterile
Supplier:  HARTMANN PAUL
Certificates
undefined
Peha-Soft® Nitrile Nitrile Gloves, Sterile
Supplier:  HARTMANN PAUL
Certificates
Specifications
  • Catalog No:
  • HART9422131
  • HART9422111
  • HART9422121
  • Ambidextrous / hand-specific:
  • Ambidextrous
  • Ambidextrous
  • Ambidextrous
  • Disposable / reusable:
  • Disposable
  • Disposable
  • Disposable
  • Latex / latex-free:
  • Latex-free
  • Latex-free
  • Latex-free
  • Powdered / powder-free:
  • Powder-free
  • Powder-free
  • Powder-free
  • Finish:
  • Textured fingertips
  • Textured fingertips
  • Textured fingertips
  • Cuff style:
  • Elastic
  • Elastic
  • Elastic
  • EN norm:
  • EN 374
  • EN 374
  • EN 374
  • Food contact approved:
  • +
  • +
  • +