Order Entry
Czech Republic
ContactUsLinkComponent
 
Wipes for delicate tasks, DuPont™ Sontara® Multipurpose Wipe
  CONTMP1213BG
 :  CONTEC
undefined
Wipes for delicate tasks, DuPont™ Sontara® Multipurpose Wipe
  CONTMP1213BG
 :  CONTEC

 

  • Packed:
    50 wipes/bag; 18 bags/case
  • Colour:
    Blue
  • Description:
    Quarterfolded wipes
  • L×W:
    305×330 mm
  • Seq:
    01
  • Pk:
    900

 

 

These wipes made from creped spunlaced polyester/cellulose fabric offer outstanding strength and sorbency. Ideal for the removal of thick liquids, greases and dirt.

  • Exceptional performance at an economical price
  • Blue wipes are creped to improve softness
  • White textured wipes scrub, trap and remove contaminants efficiently

Applications: Cleaning in less critical environments, cleaning of glass and smooth surfaces, food processing equipment sanitation and maintenance, and equipment, tool, production line and facility maintenance outside of controlled environments.

 : *The product(s) on this page are outside the scope of the EU Directive 2019/904