Order Entry
Czech Republic
ContactUsLinkComponent
 
Dual gel electrophoresis system, Owl™ P82
  720-2856
undefined
Dual gel electrophoresis system, Owl™ P82
  720-2856

 

  • Description:
    Dual gel electrophoresis system, Owl™ P82
  • Seq:
    01
  • Model:
    Owl P82
  • Gel size W×H:
    124×170 mm
  • Unit W×H×D:
    124×170×152 mm
  • Running buffer volume:
    150 ml (upper chamber 150 ml, lower chamber 300 ml)
  • Pk:
    1