ContactUsLinkComponent
Combs for horizontal gel electrophoresis system, Owl™ D4
Combs for horizontal gel electrophoresis system, Owl™ D4
Catalog # 730-0231
undefined
Combs for horizontal gel electrophoresis system, Owl™ D4
Catalog # 730-0231
Specifications
  • Description:
    Comb, micro-well 17- tooth,1,5 mm thick
  • Seq:
    02
  • Pk:
    1