Order Entry
Czech Republic
ContactUsLinkComponent
1,1-Dicyano-6-(4-N,N-dimethylaminophenyl)-1,3,5-hexatriene) (DCDAPH) fluorescent dye
1,1-Dicyano-6-(4-N,N-dimethylaminophenyl)-1,3,5-hexatriene) (DCDAPH) fluorescent dye
Catalog # 80030.
Supplier:  Biotium
CAS Number:  
1,1-Dicyano-6-(4-N,N-dimethylaminophenyl)-1,3,5-hexatriene) (DCDAPH) fluorescent dye
Catalog # 80030.
Supplier:  Biotium
CAS Number:  

Specifications

  • Pk:
    5 mg
  • Seq:
    2210

Specifications

About this item

 :
 :249.31 g/mol
Storage Temperature:Freezer
Formula:C₁₆H₁₅N₃