Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 Kč
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Czech Republic
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
DiIC₁₂(3) perchlorate
DiIC₁₂(3) perchlorate ≥95% (by TLC) fluorescent dye
DiIC₁₂(3) perchlorate ≥95% (by TLC) fluorescent dye
DiIC₁₂(3) perchlorate ≥95% (by TLC) fluorescent dye
Catalog # 734-1436
Supplier:
Corning
CAS Number:
Certificates
Expand image
DiIC₁₂(3) perchlorate ≥95% (by TLC) fluorescent dye
Catalog # 734-1436
Supplier:
Corning
CAS Number:
Certificates
Specifications
Pk:
100 mg
Seq:
2440
Specifications
About this item
Product Details
Documentation
:
:
765.5577 g/mol
CAS Number:
75664-01-6
Formula:
C₄₇H₇₃N₂O₄Cl
Product Details
Documentation