Zadání objednávky
Czech Republic
ContactUsLinkComponent
Phenol red, sodium salt
Phenol red, sodium salt
Catalog # P5648-10GR
Supplier:  Biowest
CAS Number:  
Phenol red, sodium salt
Catalog # P5648-10GR
Supplier:  Biowest
CAS Number:  
Specifications
  • Bal.:
    10 g
  • Popis:
    Phenol red, sodium salt
Specifications
About this item
Phenol red is used as a pH indicator in cell culture. Its colour exhibits a gradual transition from yellow to red over the pH range 6,8 to 8,2. Above pH 8,2, phenol red turns a bright pink (fuchsia) colour.