Comparator kits, multiparameter for water testing, Comparator 2000+
Catalog # 705-0787
Supplier: Lovibond
Specifications
- Range:Chloride: 0 - 5000 mg/l Cl
Chlorine: 0,02 - 0,3 mg/l Cl₂
Chlorine: 0,2 - 4 mg/l Cl₂
Fluoride: 0 - 1,6 mg/l F
Hardness: 0 - 500 mg/l CaCO₃
Hazen colour: 10 - 90 mg Pt/l
pH value: 6 - 8,4 - Description:Drinking water
- Seq:02
- Pk:1 KIT
- Type:AF 357
Specifications
About this item
Lovibond® Comparator system 2000+ is an extremely versatile, modular system for testing water. The battery operated portable lighting unit can be integrated to give standardised lighting conditions to enhance reproducibility.
- Compact and portable
- Simple to use yet uncompromising in terms of precision and reproducibility of results
Delivery: Comparator test kits are supplied as a complete system in a sturdy plastic case. Together with the Comparator 2000+ and test discs, each kit includes all the necessary cells, accessories and Lovibond® tablet reagents (for 100 measurements). The AF368 is not supplied with reagents.