Industrial food scales, Valor® 2000
: OHAUS
Waterproof compact scales designed for the food industry. Features include keypad with two mechanical switches on V22PWE models and four on V22XWE models, adjustable feet, level bubble and cover with spring to protect power connector of the scale from ingress of moisture, especially when the scale is washed down. Dual LED display back and front enable two operators to work with the Valor® 2000 at the same time. Choice of scale with plastic or stainless steel housing, the models with stainless steel housing also have a check weighing application mode.
- Stabilisation times of 0,5 s
- Flow Thru™ design protects scales from water damage - water drains away ensuring the scale remains clean and operational
- Thick polycarbonate keypad is impact and puncture resistant
- Easy to clean and sanitary design support HACCP certified systems
- Carrying handle and rechargeable lead acid battery with up to 50 hours of continuous usage time, allowing for portable operation
Frequently Bought Together