Order Entry
Czech Republic
ContactUsLinkComponent
 
Wiping paper, Tork Reflex™
  115-3120
 :  Essity
undefined
Wiping paper, Tork Reflex™
  115-3120
 :  Essity

 

  • For:
    M4
  • Packed:
    6 rolls of 150,8 m
  • Colour:
    White
  • Description:
    Wiping paper roll, centrefeed, 2 ply, 450 sheets
  • L×W:
    150,8 m × 194 mm
  • Pk:
    6 Roll

 

 

This multi-purpose paper is ideal for most wiping tasks, especially for mopping up spills.

  • Tork Easy Handling™: packaging is easy to carry, to open, to flatten and to carry away
  • Soft and therefore also suitable for hand wiping
  • Can be used with M4 dispensers (Centrefeed Reflex Roll System)